Anti-SND1/P100 Mouse Monoclonal Antibody [clone: 13B719]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030641EA 3930 DKK
USBI030641
Anti-SND1/P100 Mouse Monoclonal Antibody [clone: 13B719]
Antistoffer
Type: Primary
Antigen: SND1/P100
Clonality: Monoclonal
Clone: 13B719
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human