Anti-SNCG Mouse Monoclonal Antibody [clone: 13B718]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030640EA 2910 DKK
USBI030640
Anti-SNCG Mouse Monoclonal Antibody [clone: 13B718]
Antistoffer
Type: Primary
Antigen: SNCG (synuclein, gamma (breast cancer-specific protein 1))
Clonality: Monoclonal
Clone: 13B718
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR