Anti-SMAD5 Mouse Monoclonal Antibody [clone: 13B710]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030632EA 3080 DKK
USBI030632
Anti-SMAD5 Mouse Monoclonal Antibody [clone: 13B710]
Antistoffer
Type: Primary
Antigen: SMAD5 (SMAD family member 5)
Clonality: Monoclonal
Clone: 13B710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR