Anti-SHH Mouse Monoclonal Antibody [clone: 13B701]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030623EA 3270 DKK
USBI030623
Anti-SHH Mouse Monoclonal Antibody [clone: 13B701]
Antistoffer
Type: Primary
Antigen: SHH (sonic hedgehog)
Clonality: Monoclonal
Clone: 13B701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR