Anti-SARS, E2 Mouse Monoclonal Antibody [clone: 13B694]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030616EA 2300 DKK
USBI030616
Anti-SARS, E2 Mouse Monoclonal Antibody [clone: 13B694]
Antistoffer
Type: Primary
Antigen: SARS, E2
Clonality: Monoclonal
Clone: 13B694
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR