Anti-S100A10 Mouse Monoclonal Antibody [clone: 13B692]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030614EA 3780 DKK
USBI030614
Anti-S100A10 Mouse Monoclonal Antibody [clone: 13B692]
Antistoffer
Type: Primary
Antigen: S100A10 (S100 calcium binding protein A10)
Clonality: Monoclonal
Clone: 13B692
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR