Anti-RTN3 Mouse Monoclonal Antibody [clone: 13B686]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030608EA 2720 DKK
USBI030608
Anti-RTN3 Mouse Monoclonal Antibody [clone: 13B686]
Antistoffer
Type: Primary
Antigen: RTN3 (reticulon 3)
Clonality: Monoclonal
Clone: 13B686
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR