Anti-RPS6KA3 Mouse Monoclonal Antibody [clone: 13B685]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030607EA 4410 DKK
USBI030607
Anti-RPS6KA3 Mouse Monoclonal Antibody [clone: 13B685]
Antistoffer
Type: Primary
Antigen: RPS6KA3 (ribosomal protein S6 kinase, 90kDa, polypeptide 3)
Clonality: Monoclonal
Clone: 13B685
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR