Anti-ROCK1 Mouse Monoclonal Antibody [clone: 13B679]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030601EA 3270 DKK
USBI030601
Anti-ROCK1 Mouse Monoclonal Antibody [clone: 13B679]
Antistoffer
Type: Primary
Antigen: ROCK1 (Rho-associated, coiled-coil containing protein kinase 1)
Clonality: Monoclonal
Clone: 13B679
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR