Anti-REG1A Mouse Monoclonal Antibody [clone: 13B674]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030596EA 3930 DKK
USBI030596
Anti-REG1A Mouse Monoclonal Antibody [clone: 13B674]
Antistoffer
Type: Primary
Antigen: REG1A (regenerating islet-derived 1 alpha)
Clonality: Monoclonal
Clone: 13B674
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR