Anti-PTPN11 Mouse Monoclonal Antibody [clone: 13B658]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030580EA 3270 DKK
USBI030580
Anti-PTPN11 Mouse Monoclonal Antibody [clone: 13B658]
Antistoffer
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: Monoclonal
Clone: 13B658
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR