Anti-PTH Mouse Monoclonal Antibody [clone: 13B654]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030576EA 3080 DKK
USBI030576
Anti-PTH Mouse Monoclonal Antibody [clone: 13B654]
Antistoffer
Type: Primary
Antigen: PTH (parathyroid hormone)
Clonality: Monoclonal
Clone: 13B654
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR