Anti-PSMB8 Mouse Monoclonal Antibody [clone: 13B652]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030574EA 4410 DKK
USBI030574
Anti-PSMB8 Mouse Monoclonal Antibody [clone: 13B652]
Antistoffer
Type: Primary
Antigen: PSMB8 (proteasome (prosome, macropain) subunit, beta type, 8)
Clonality: Monoclonal
Clone: 13B652
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR