Anti-PSIP1 Mouse Monoclonal Antibody [clone: 13B651]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030573EA 4230 DKK
USBI030573
Anti-PSIP1 Mouse Monoclonal Antibody [clone: 13B651]
Antistoffer
Type: Primary
Antigen: PSIP1 (PC4 and SFRS1 interacting protein 1)
Clonality: Monoclonal
Clone: 13B651
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR