Anti-PRMT6 Mouse Monoclonal Antibody [clone: 13B646]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030568EA 3080 DKK
USBI030568
Anti-PRMT6 Mouse Monoclonal Antibody [clone: 13B646]
Antistoffer
Type: Primary
Antigen: PRMT6 (protein arginine methyltransferase 6)
Clonality: Monoclonal
Clone: 13B646
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR