Anti-PIK3R1 Mouse Monoclonal Antibody [clone: 13B625]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030547EA 3270 DKK
USBI030547
Anti-PIK3R1 Mouse Monoclonal Antibody [clone: 13B625]
Antistoffer
Type: Primary
Antigen: PIK3R1 (phosphoinositide-3-kinase, regulatory subunit 1 (alpha))
Clonality: Monoclonal
Clone: 13B625
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR