Anti-F2RL3 Mouse Monoclonal Antibody [clone: 13B602]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030524EA 3330 DKK
USBI030524
Anti-F2RL3 Mouse Monoclonal Antibody [clone: 13B602]
Antistoffer
Type: Primary
Antigen: F2RL3 (coagulation factor II (thrombin) receptor-like 3)
Clonality: Monoclonal
Clone: 13B602
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR