Anti-OTX2 Mouse Monoclonal Antibody [clone: 13B591]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030513EA 3350 DKK
USBI030513
Anti-OTX2 Mouse Monoclonal Antibody [clone: 13B591]
Antistoffer
Type: Primary
Antigen: OTX2 (orthodenticle homeobox 2)
Clonality: Monoclonal
Clone: 13B591
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR