Anti-NCOR1 Mouse Monoclonal Antibody [clone: 13B558]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030480EA 3330 DKK
USBI030480
Anti-NCOR1 Mouse Monoclonal Antibody [clone: 13B558]
Antistoffer
Type: Primary
Antigen: NCOR1 (nuclear receptor corepressor 1)
Clonality: Monoclonal
Clone: 13B558
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR