Anti-NACC1 Mouse Monoclonal Antibody [clone: 13B554]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030476EA 3930 DKK
USBI030476
Anti-NACC1 Mouse Monoclonal Antibody [clone: 13B554]
Antistoffer
Type: Primary
Antigen: NACC1 (nucleus accumbens associated 1, BEN and BTB (POZ) domain containing)
Clonality: Monoclonal
Clone: 13B554
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR