Anti-MUM1 Mouse Monoclonal Antibody [clone: 13B542]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030464EA 3080 DKK
USBI030464
Anti-MUM1 Mouse Monoclonal Antibody [clone: 13B542]
Antistoffer
Type: Primary
Antigen: MUM1 (melanoma associated antigen (mutated) 1)
Clonality: Monoclonal
Clone: 13B542
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR