Anti-MSI2 Mouse Monoclonal Antibody [clone: 13B538]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030460EA 3270 DKK
USBI030460
Anti-MSI2 Mouse Monoclonal Antibody [clone: 13B538]
Antistoffer
Type: Primary
Antigen: MSI2 (musashi RNA-binding protein 2)
Clonality: Monoclonal
Clone: 13B538
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR