Anti-MSI1 Mouse Monoclonal Antibody [clone: 13B537]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030459EA 3270 DKK
USBI030459
Anti-MSI1 Mouse Monoclonal Antibody [clone: 13B537]
Antistoffer
Type: Primary
Antigen: MSI1 (musashi RNA-binding protein 1)
Clonality: Monoclonal
Clone: 13B537
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR