Anti-Mouse TUG Mouse Monoclonal Antibody [clone: 13B529]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030451EA 2910 DKK
USBI030451
Anti-Mouse TUG Mouse Monoclonal Antibody [clone: 13B529]
Antistoffer
Type: Primary
Antigen: Mouse TUG
Clonality: Monoclonal
Clone: 13B529
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR