Anti-MMP3 Mouse Monoclonal Antibody [clone: 13B526]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030448EA 3270 DKK
USBI030448
Anti-MMP3 Mouse Monoclonal Antibody [clone: 13B526]
Antistoffer
Type: Primary
Antigen: MMP3 (matrix metallopeptidase 3 (stromelysin 1, progelatinase))
Clonality: Monoclonal
Clone: 13B526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR