Anti-MESP1 Mouse Monoclonal Antibody [clone: 13B520]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030442EA 3080 DKK
USBI030442
Anti-MESP1 Mouse Monoclonal Antibody [clone: 13B520]
Antistoffer
Type: Primary
Antigen: MESP1 (mesoderm posterior 1 homolog (mouse))
Clonality: Monoclonal
Clone: 13B520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR