Anti-MELK Mouse Monoclonal Antibody [clone: 13B518]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030440EA 3270 DKK
USBI030440
Anti-MELK Mouse Monoclonal Antibody [clone: 13B518]
Antistoffer
Type: Primary
Antigen: MELK (maternal embryonic leucine zipper kinase)
Clonality: Monoclonal
Clone: 13B518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR