Anti-CCL2 Mouse Monoclonal Antibody [clone: 13B515]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030437EA 2840 DKK
USBI030437
Anti-CCL2 Mouse Monoclonal Antibody [clone: 13B515]
Antistoffer
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: Monoclonal
Clone: 13B515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR