Anti-MCM2 Mouse Monoclonal Antibody [clone: 13B513]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030435EA 4410 DKK
USBI030435
Anti-MCM2 Mouse Monoclonal Antibody [clone: 13B513]
Antistoffer
Type: Primary
Antigen: MCM2 (minichromosome maintenance complex component 2)
Clonality: Monoclonal
Clone: 13B513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR