Anti-MAP3K5 Mouse Monoclonal Antibody [clone: 13B500]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030422EA 3270 DKK
USBI030422
Anti-MAP3K5 Mouse Monoclonal Antibody [clone: 13B500]
Antistoffer
Type: Primary
Antigen: MAP3K5 (mitogen-activated protein kinase kinase kinase 5)
Clonality: Monoclonal
Clone: 13B500
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR