Anti-KRT19 Mouse Monoclonal Antibody [clone: 13B463]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030385EA 4410 DKK
USBI030385
Anti-KRT19 Mouse Monoclonal Antibody [clone: 13B463]
Antistoffer
Type: Primary
Antigen: KRT19 (keratin 19)
Clonality: Monoclonal
Clone: 13B463
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human