Anti-Prostate Specific Antigen Mouse Monoclonal Antibody [clone: 13B460]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030382EA 3270 DKK
USBI030382
Anti-Prostate Specific Antigen Mouse Monoclonal Antibody [clone: 13B460]
Antistoffer
Type: Primary
Antigen: Prostate Specific Antigen
Clonality: Monoclonal
Clone: 13B460
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human