Anti-KLHL22 Mouse Monoclonal Antibody [clone: 13B458]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030380EA 4030 DKK
USBI030380
Anti-KLHL22 Mouse Monoclonal Antibody [clone: 13B458]
Antistoffer
Type: Primary
Antigen: KLHL22 (kelch-like family member 22)
Clonality: Monoclonal
Clone: 13B458
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human