Anti-KEAP1 Mouse Monoclonal Antibody [clone: 13B441]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030363EA 4410 DKK
USBI030363
Anti-KEAP1 Mouse Monoclonal Antibody [clone: 13B441]
Antistoffer
Type: Primary
Antigen: KEAP1 (kelch-like ECH-associated protein 1)
Clonality: Monoclonal
Clone: 13B441
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse