Anti-JAK3 Mouse Monoclonal Antibody [clone: 13B433]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030355EA 3780 DKK
USBI030355
Anti-JAK3 Mouse Monoclonal Antibody [clone: 13B433]
Antistoffer
Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: Monoclonal
Clone: 13B433
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse