Anti-JAK2 Mouse Monoclonal Antibody [clone: 13B432]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030354EA 3180 DKK
USBI030354
Anti-JAK2 Mouse Monoclonal Antibody [clone: 13B432]
Antistoffer
Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: Monoclonal
Clone: 13B432
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR