Anti-IFNG Mouse Monoclonal Antibody [clone: 13B393]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030315EA 2840 DKK
USBI030315
Anti-IFNG Mouse Monoclonal Antibody [clone: 13B393]
Antistoffer
Type: Primary
Antigen: IFNG (interferon, gamma)
Clonality: Monoclonal
Clone: 13B393
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR