Anti-ICAM1 Mouse Monoclonal Antibody [clone: 13B392]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030314EA 3080 DKK
USBI030314
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 13B392]
Antistoffer
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: Monoclonal
Clone: 13B392
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR