Anti-HSPA5 Mouse Monoclonal Antibody [clone: 13B390]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030312EA 3080 DKK
USBI030312
Anti-HSPA5 Mouse Monoclonal Antibody [clone: 13B390]
Antistoffer
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: Monoclonal
Clone: 13B390
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR