Anti-HDAC3 Mouse Monoclonal Antibody [clone: 13B371]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030293EA 3330 DKK
USBI030293
Anti-HDAC3 Mouse Monoclonal Antibody [clone: 13B371]
Antistoffer
Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: Monoclonal
Clone: 13B371
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR