Anti-HCK Mouse Monoclonal Antibody [clone: 13B369]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030291EA 3330 DKK
USBI030291
Anti-HCK Mouse Monoclonal Antibody [clone: 13B369]
Antistoffer
Type: Primary
Antigen: HCK (hemopoietic cell kinase)
Clonality: Monoclonal
Clone: 13B369
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR