Anti-GRIA3 Mouse Monoclonal Antibody [clone: 13B353]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030275EA 3270 DKK
USBI030275
Anti-GRIA3 Mouse Monoclonal Antibody [clone: 13B353]
Antistoffer
Type: Primary
Antigen: GRIA3 (glutamate receptor, ionotropic, AMPA 3)
Clonality: Monoclonal
Clone: 13B353
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human