Anti-DLGAP1 Mouse Monoclonal Antibody [clone: 13B346]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030268EA 4410 DKK
USBI030268
Anti-DLGAP1 Mouse Monoclonal Antibody [clone: 13B346]
Antistoffer
Type: Primary
Antigen: DLGAP1 (discs, large (Drosophila) homolog-associated protein 1)
Clonality: Monoclonal
Clone: 13B346
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR