Anti-GAPDH Mouse Monoclonal Antibody [clone: 13B335]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030257EA 2920 DKK
USBI030257
Anti-GAPDH Mouse Monoclonal Antibody [clone: 13B335]
Antistoffer
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: Monoclonal
Clone: 13B335
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human