Anti-GAB1 Mouse Monoclonal Antibody [clone: 13B333]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030255EA 4410 DKK
USBI030255
Anti-GAB1 Mouse Monoclonal Antibody [clone: 13B333]
Antistoffer
Type: Primary
Antigen: GAB1 (GRB2-associated binding protein 1)
Clonality: Monoclonal
Clone: 13B333
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human