Anti-G6PD Mouse Monoclonal Antibody [clone: 13B331]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030253EA 4410 DKK
USBI030253
Anti-G6PD Mouse Monoclonal Antibody [clone: 13B331]
Antistoffer
Type: Primary
Antigen: G6PD (glucose-6-phosphate dehydrogenase)
Clonality: Monoclonal
Clone: 13B331
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human