Anti-FCER2 Mouse Monoclonal Antibody [clone: 13B295]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030217EA 3180 DKK
USBI030217
Anti-FCER2 Mouse Monoclonal Antibody [clone: 13B295]
Antistoffer
Type: Primary
Antigen: FCER2 (Fc fragment of IgE, low affinity II, receptor for (CD23))
Clonality: Monoclonal
Clone: 13B295
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR