Anti-PTK2 Mouse Monoclonal Antibody [clone: 13B287]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030209EA 2470 DKK
USBI030209
Anti-PTK2 Mouse Monoclonal Antibody [clone: 13B287]
Antistoffer
Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: Monoclonal
Clone: 13B287
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR