Anti-EIF4B Mouse Monoclonal Antibody [clone: 13B282]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030204EA 4410 DKK
USBI030204
Anti-EIF4B Mouse Monoclonal Antibody [clone: 13B282]
Antistoffer
Type: Primary
Antigen: EIF4B (eukaryotic translation initiation factor 4B)
Clonality: Monoclonal
Clone: 13B282
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR