Anti-MAPK1 Mouse Monoclonal Antibody [clone: 13B270]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030192EA 2840 DKK
USBI030192
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 13B270]
Antistoffer
Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Monoclonal
Clone: 13B270
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR